Merck
  • Binding of longer Aβ to transmembrane domain 1 of presenilin 1 impacts on Aβ42 generation.
Binding of longer Aβ to transmembrane domain 1 of presenilin 1 impacts on Aβ42 generation.
Molecular neurodegeneration (2014-01-15)
Yu Ohki, Naoaki Shimada, Aya Tominaga, Satoko Osawa, Takuya Higo, Satoshi Yokoshima, Tohru Fukuyama, Taisuke Tomita, Takeshi Iwatsubo
ABSTRACT
Amyloid-β peptide ending at 42nd residue (Aβ42) is believed as a pathogenic peptide for Alzheimer disease. Although γ-secretase is a responsible protease to generate Aβ through a processive cleavage, the proteolytic mechanism of γ-secretase at molecular level is poorly understood. We found that the transmembrane domain (TMD) 1 of presenilin (PS) 1, a catalytic subunit for the γ-secretase, as a key modulatory domain for Aβ42 production. Aβ42-lowering and -raising γ-secretase modulators (GSMs) directly targeted TMD1 of PS1 and affected its structure. A point mutation in TMD1 caused an aberrant secretion of longer Aβ species including Aβ45 that are the precursor of Aβ42. We further found that the helical surface of TMD1 is involved in the binding of Aβ45/48 and that the binding was altered by GSMs as well as TMD1 mutation. Binding between PS1 TMD1 and longer Aβ is critical for Aβ42 production.
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution