Merck
  • Tetratricopeptide repeat domain 9A modulates anxiety-like behavior in female mice.
Tetratricopeptide repeat domain 9A modulates anxiety-like behavior in female mice.
Scientific reports (2016-11-22)
Lee Wei Lim, Smeeta Shrestha, Yu Zuan Or, Shawn Zheng Kai Tan, Hwa Hwa Chung, Yang Sun, Chew Leng Lim, Sharafuddin Khairuddin, Thomas Lufkin, Valerie Chun Ling Lin
ABSTRACT
Tetratricopeptide repeat domain 9A (TTC9A) expression is abundantly expressed in the brain. Previous studies in TTC9A knockout (TTC9A
Sigma-Aldrich
5-Methyl-2-hexanol, 98%