Merck
  • Tetramethylammonium dichloroiodate: an efficient and environmentally friendly iodination reagent for iodination of aromatic compounds under mild and solvent-free conditions.
Tetramethylammonium dichloroiodate: an efficient and environmentally friendly iodination reagent for iodination of aromatic compounds under mild and solvent-free conditions.
The Journal of organic chemistry (2002-11-26)
Abdol R Hajipour, Marty Arbabian, Arnold E Ruoho
ABSTRACT
Tetramethylammonium dichloroiodate (1, TMADCI) as a mild and efficient iodination reagent was prepared. Iodination of different aromatic compounds with this reagent takes place fast and with high yields under solvent-free conditions.
Sigma-Aldrich
Tetramethylammonium iodide, 99%