Merck
  • Perylene bisimide dyes as versatile building blocks for functional supramolecular architectures.
Perylene bisimide dyes as versatile building blocks for functional supramolecular architectures.
Chemical communications (Cambridge, England) (2004-07-21)
Frank Würthner
ABSTRACT
Perylene bisimide dyes and their organization into supramolecular architectures through hydrogen-bonding, metal ion coordination and pi-pi-stacking is discussed; further self-assembly leading to nano- and meso-scopic structures and liquid-crystalline compounds is also addressed.
Sigma-Aldrich
N,N′-Dipentyl-3,4,9,10-perylenedicarboximide, 98%
Sigma-Aldrich
N,N′-Dioctyl-3,4,9,10-perylenedicarboximide, 98%