Merck
  • Dysregulation of FMRP/mTOR Signaling Cascade in Hypoxic-Ischemic Injury of Premature Human Brain.
Dysregulation of FMRP/mTOR Signaling Cascade in Hypoxic-Ischemic Injury of Premature Human Brain.
Journal of child neurology (2015-08-05)
Mirna Lechpammer, Pia Wintermark, Katherine M Merry, Michele C Jackson, Lauren L Jantzie, Frances E Jensen
ABSTRACT
In this study the authors investigated whether dysregulation of the fragile X mental retardation protein and mammalian target of rapamycin signaling cascade can have a role in the pathogenesis of encephalopathy of prematurity following perinatal hypoxia-ischemia. The authors examined the brain tissue of newborns with encephalopathy and compared it to age-matched controls with normal brain development and adults. In normal controls, the fragile X mental retardation protein expression in cortical gray matter spiked 4-fold during 36-39 gestational weeks compared to the adult, with a concomitant suppression of p70S6K and S6. In encephalopathy cases, the developmental spike of fragile X mental retardation protein was not observed, and fragile X mental retardation protein levels remained significantly lower than in normal controls. Importantly, this fragile X mental retardation protein downregulation was followed by a significant overexpression of p70S6K and S6. These novel findings thus suggest that premature hypoxic-ischemic brain injury can affect the fragile X mental retardation protein/mammalian target of rapamycin pathway, as otherwise observed in inherited syndromes of cognitive disability and autism spectrum disorders.
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-mGluR5 Antibody, clone N75/33, clone N75/33, from mouse