Merck
Photovoltaic Performance of PbS Quantum Dots Treated with Metal Salts.
ACS nano (2016-02-26)
Dong-Kyun Ko, Andrea Maurano, Su Kyung Suh, Donghun Kim, Gyu Weon Hwang, Jeffrey C Grossman, Vladimir Bulović, Moungi G Bawendi
ABSTRACT
Recent advances in quantum dot surface passivation have led to a rapid development of high-efficiency solar cells. Another critical element for achieving efficient power conversion is the charge neutrality of quantum dots, as charge imbalances induce electronic states inside the energy gap. Here we investigate how the simultaneous introduction of metal cations and halide anions modifies the charge balance and enhances the solar cell efficiency. The addition of metal salts between QD deposition and ligand exchange with 1,3-BDT results in an increase in the short-circuit current and fill factor, accompanied by a distinct reduction in a crossover between light and dark current density-voltage characteristics.
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
1,3-Benzenedithiol, 99%