- Conversion of Xylose to Furfuryl Alcohol and 2-Methylfuran in a Continuous Fixed-Bed Reactor.
Conversion of Xylose to Furfuryl Alcohol and 2-Methylfuran in a Continuous Fixed-Bed Reactor.
ChemSusChem (2016-04-28)
Jinglei Cui, Jingjing Tan, Xiaojing Cui, Yulei Zhu, Tiansheng Deng, Guoqiang Ding, Yongwang Li
PMID27120138
ABSTRACT
An efficient process was designed for the synthesis of furfuryl alcohol and 2-methylfuran from xylose using a continuous fixed-bed reactor over a catalyst combining Hβ zeolite and Cu/ZnO/Al2 O3 in γ-butyrolactone (GBL)/water as solvent. The cooperative effect of Hβ zeolite and GBL facilitated the dehydration of xylose and enhanced largely the furfural yield. The production of furfuryl alcohol and 2-methylfuran can be simply tuned by changing the hydrogenation temperature for furfural over the Cu/ZnO/Al2 O3 catalyst. The yield for furfuryl alcohol reached 87.2 % at 150 °C whereas a yield of 86.8 % was achieved for 2-methylfuran at 190 °C.