Merck
The electroneutral Na(+)-(K+)-Cl- cotransport family.
Kidney international (1996-06-01)
S C Hebert, G Gamba, M Kaplan
ABSTRACT
Recently the molecular identification of the major electroneutral sodium-potassium-chloride entry mechanisms present on apical membranes of distal nephron segments of the mammalian kidney, on basolateral membranes of many non-renal epithelial cells and on certain non-epithelial tissues has been achieved. These transporters represent a major pathway for cellular uptake of chloride critical for chloride absorptive and secretory processes and for cell volume regulation following cell shrinkage. In the mammalian kidney, these sodium-coupled chloride cotransporters represent the major target sites for clinically useful diuretics including the "loop" diuretics [furosemide (Lasix) and bumetanide (Bumex)] and thiazides (such as, chlorothiazide, hydrochlorothiazide and metolazone). Although these Na-(K)-Cl cotransporters exhibit functional and pharmacological differences, they clearly evolved from a common ancestral gene and thus form a new gene family. This information is already advancing our understanding of the evolution, structure and function of these transporters both in renal handling of sodium and in hypertension.
Sigma-Aldrich
Metolazone, ≥98% (HPLC), solid