Merck
  • The transcription factor EGR1 localizes to the nucleolus and is linked to suppression of ribosomal precursor synthesis.
The transcription factor EGR1 localizes to the nucleolus and is linked to suppression of ribosomal precursor synthesis.
PloS one (2014-05-03)
Donatella Ponti, Gian Carlo Bellenchi, Rosa Puca, Daniela Bastianelli, Marella Maroder, Giuseppe Ragona, Pascal Roussel, Marc Thiry, Dan Mercola, Antonella Calogero
ABSTRACT
EGR1 is an immediate early gene with a wide range of activities as transcription factor, spanning from regulation of cell growth to differentiation. Numerous studies show that EGR1 either promotes the proliferation of stimulated cells or suppresses the tumorigenic growth of transformed cells. Upon interaction with ARF, EGR1 is sumoylated and acquires the ability to bind to specific targets such as PTEN and in turn to regulate cell growth. ARF is mainly localized to the periphery of nucleolus where is able to negatively regulate ribosome biogenesis. Since EGR1 colocalizes with ARF under IGF-1 stimulation we asked the question of whether EGR1 also relocate to the nucleolus to interact with ARF. Here we show that EGR1 colocalizes with nucleolar markers such as fibrillarin and B23 in the presence of ARF. Western analysis of nucleolar extracts from HeLa cells was used to confirm the presence of EGR1 in the nucleolus mainly as the 100 kDa sumoylated form. We also show that the level of the ribosomal RNA precursor 47S is inversely correlated to the level of EGR1 transcripts. The EGR1 iseffective to regulate the synthesis of the 47S rRNA precursor. Then we demonstrated that EGR1 binds to the Upstream Binding Factor (UBF) leading us to hypothesize that the regulating activity of EGR1 is mediated by its interaction within the transcriptional complex of RNA polymerase I. These results confirm the presence of EGR1 in the nucleolus and point to a role for EGR1 in the control of nucleolar metabolism.
Sigma-Aldrich
Anti-Sumo-1 antibody, Rat monoclonal, clone 4D12, purified from hybridoma cell culture