Merck
  • Proton nuclear magnetic resonance studies on methylthiohydantoins, thiohydantoins, and hydantoins of amino acids.
Proton nuclear magnetic resonance studies on methylthiohydantoins, thiohydantoins, and hydantoins of amino acids.
Canadian journal of biochemistry (1977-05-01)
T Suzuki, T Tomioka, K Tuzimura
PMID884586
ABSTRACT
Proton magnetic resonance spectra of methylthiohydantoin (3-methyl-2-thiohydantoin), thiohydantoin (2-thiohydantoin), and hydantoin derivatives of amino acids were studied in dimethyl sulfoxide-d6. Their parent amino acids could be identified by the spectra. An application to the N- and C-terminal amino acid analysis of a tripeptide was examined.
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)