Merck
  • Hearing loss and hair cell death in mice given the cholesterol-chelating agent hydroxypropyl-β-cyclodextrin.
Hearing loss and hair cell death in mice given the cholesterol-chelating agent hydroxypropyl-β-cyclodextrin.
PloS one (2013-01-04)
Mark A Crumling, Liqian Liu, Paul V Thomas, Jennifer Benson, Ariane Kanicki, Lisa Kabara, Karin Hälsey, David Dolan, R Keith Duncan
ABSTRACT
Cyclodextrins are sugar compounds that are increasingly finding medicinal uses due to their ability to complex with hydrophobic molecules. One cyclodextrin in particular, 2-hydroxypropyl-β-cyclodextrin (HPβCD), is used as a carrier to solubilize lipophilic drugs and is itself being considered as a therapeutic agent for treatment of Niemann-Pick Type C disease, due to its ability to mobilize cholesterol. Results from toxicological studies suggest that HPβCD is generally safe, but a recent study has found that it causes hearing loss in cats. Whether the hearing loss occurred via death of cochlear hair cells, rendering it permanent, was unexplored. In the present study, we examined peripheral auditory function and cochlear histology in mice after subcutaneous injection of HPβCD to test for hearing loss and correlate any observed auditory deficits with histological findings. On average, auditory brainstem response thresholds were elevated at 4, 16, and 32 kHz in mice one week after treatment with 8,000 mg/kg. In severely affected mice all outer hair cells were missing in the basal half of the cochlea. In many cases, surviving hair cells in the cochlear apex exhibited abnormal punctate distribution of the motor protein prestin, suggesting long term changes to membrane composition and integrity. Mice given a lower dose of 4,000 mg/kg exhibited hearing loss only after repeated doses, but these threshold shifts were temporary. Therefore, cyclodextrin-induced hearing loss was complex, involving cell death and other more subtle influences on cochlear physiology.
Product Description
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®