Merck
  • USP17-mediated deubiquitination and stabilization of HDAC2 in cigarette smoke extract-induced inflammation.
USP17-mediated deubiquitination and stabilization of HDAC2 in cigarette smoke extract-induced inflammation.
International journal of clinical and experimental pathology (2015-12-01)
Huihui Song, Lianqin Tao, Chen Chen, Lina Pan, Jimin Hao, Yingmeng Ni, Dan Li, Bin Li, Guochao Shi
ABSTRACT
Histone deacetylase HDAC2 regulates genes transcription via removing the acetyl group from histones. Glucocorticoids, the most potent anti-inflammatory treatment available for inflammatory diseases, inhibit the expression of inflammatory genes by recruiting HDAC2 to activated genes. In the lungs of patients who smoke and have chronic obstructive pulmonary disease (COPD) or asthma, glucocorticoids are not effective enough to suppress airway inflammation, which is so called "glucocorticoid resistance", due to decreased HDAC2 level caused by cigarette smoke. We report that the ubiquitin-specific protease USP17 interacts with HDAC2. USP17 deubiquitinates and stabilizes the protein level of HDAC2. In cigarette smoke extract-exposed airway epithelial cells and macrophages, HDAC2 is excessively ubiquitinated and degraded in the proteasome attributed to low expression of USP17. Furthermore, over-expression of USP17 blocks the destruction of HDAC2 induced by cigarette smoke extract. These results provide a new insight into the mechanisms of glucocorticoid resistance in airway inflammatory disease. Small molecules which can specifically induce the expression of USP17 might be useful in reversing glucocorticoid resistance.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)