Merck
  • Clinical significance and therapeutic potential of prostate cancer antigen-1/ALKBH3 in human renal cell carcinoma.
Clinical significance and therapeutic potential of prostate cancer antigen-1/ALKBH3 in human renal cell carcinoma.
Oncology reports (2015-06-03)
Kiyohiko Hotta, Masayuki Sho, Kiyohide Fujimoto, Keiji Shimada, Ichiro Yamato, Satoshi Anai, Hiroshi Harada, Kazutake Tsujikawa, Noboru Konishi, Nobuo Shinohara, Yoshiyuki Nakajima
ABSTRACT
Prostate cancer antigen-1 (PCA-1)/ALKBH3 has been recently identified in human prostate cancer and its expression is correlated with disease progression and prognosis. However, the precise role and function of PCA-1/ALKBH3 in human malignancies are largely unknown. In the present study, we investigated the clinical significance and therapeutic potential of PCA-1/ALKBH3 in renal cell carcinoma (RCC). PCA-1/ALKBH3 expression was examined by immunohistochemistry in 101 RCC patients who underwent radical or partial nephrectomy. Its expression was positively correlated with advanced pathological T- and M-factors and TNM stage (T, P<0.05; M, P<0.01; TNM, P<0.01, respectively). In the prognostic analysis, PCA-1/ALKBH3-negative patients with RCC had a significantly better prognosis than PCA-1/ALKBH3-positive patients (5-year survival rate, 92.9 vs. 75.9%, respectively; P<0.05). Next, the therapeutic potential of targeting PCA-1/ALKBH3 was further evaluated by small interfering RNA method using a human RCC cell line (CAKI-1). We found that PCA-1/ALKBH3 knockdown significantly inhibited the growth of CAKI-1 cells compared with the control (P<0.001). Furthermore, knockdown of PCA-1 induced apoptosis in CAKI-1 cells, as assessed by poly(ADP-ribose) polymerase-cleavage assays. We demonstrated for the first time that PCA-1/ALKBH3 expression has a significant prognostic impact on patient prognosis in RCC. Furthermore, its knockdown has a therapeutic efficacy on RCC. Taken together, both our clinical and experimental data strongly suggest that PCA-1/ALKBH3 may be functionally important and a novel molecular target for human RCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human ALKBH3
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)