Merck
  • Kinetics of lymphocyte subpopulations in allogeneic grafted scales of ginbuna crucian carp.
Kinetics of lymphocyte subpopulations in allogeneic grafted scales of ginbuna crucian carp.
Developmental and comparative immunology (2015-04-29)
Yasuhiro Shibasaki, Yuta Matsuura, Hideaki Toda, Nozomi Imabayashi, Tatsuyuki Nishino, Kosuke Uzumaki, Chihiro Hatanaka, Takeshi Yabu, Tadaaki Moritomo, Teruyuki Nakanishi
ABSTRACT
In mammals the rejection of allografts is primarily accomplished by cell-mediated immunity including T cells. Recently, considerable studies reveal the existence of helper and cytotoxic T cell subsets in fish. Here we investigate the kinetics of CD4(+) and CD8α(+) T cells along with sIgM(+) cells and phagocytic cells in an allogeneic scale graft model using ginbuna crucian carp for understanding the mechanisms of cell-mediated immune response. The results showed that CD4(+) T cells first infiltrated into allogeneic scales followed by CD8α(+) and sIgM(+) cells, and finally phagocytic cells appeared in the graft. Furthermore, most of the CD8α(+) T cells appeared on the border of the allografted scales at the time of rejection. These results suggest that T cells play crucial roles and work together with other cell types for completion of allograft rejection.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)