Merck
  • One-pot sonochemical synthesis of ferrocenyl derivatives via a three-component reaction in aqueous media.
One-pot sonochemical synthesis of ferrocenyl derivatives via a three-component reaction in aqueous media.
Ultrasonics sonochemistry (2015-07-19)
Luca Cappelletti, Luca Vaghi, Laura Rinaldi, Laura Rotolo, Giovanni Palmisano, Giancarlo Cravotto, Andrea Penoni
ABSTRACT
An ultrasound-assisted three-component, one-pot domino reaction with ferrocenecarboxaldehyde is herein reported. The sequence of reactions entails the allylindation and dehydrative alkylation of stabilized C-nucleophiles (e.g. electron-rich-(hetero)aromatics and stabilized enols) and N-nucleophiles (e.g. azoles). Sonochemical reactions have been performed in three different high-intensity reactors: a bath (20.3 kHz, 60 W), as well as two cup horns working at 19.9 kHz (75 W) and 300.5 kHz (70 W) giving a library of 18 new ferrocenyl derivatives.
Sigma-Aldrich
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%