Merck
  • FilGAP, a Rho-ROCK-regulated GAP for Rac, controls adherens junctions in MDCK cells.
FilGAP, a Rho-ROCK-regulated GAP for Rac, controls adherens junctions in MDCK cells.
Journal of cell science (2015-04-25)
Shinichiro Nakahara, Koji Tsutsumi, Takuya Zuinen, Yasutaka Ohta
ABSTRACT
Rho family small GTPases are essential for the formation of adherens junctions in epithelial cells. Here, we found that FilGAP (also known as ARHGAP24), a Rac-specific Rho GTPase-activating protein, promoted the formation of adherens junctions in Madin-Darby canine kidney (MDCK) cells. Knockdown of FilGAP by siRNA stimulated the disassembly and migration of MDCK cells induced by hepatocyte growth factor (HGF). By contrast, forced expression of FilGAP induced accumulation of E-cadherin at adherens junctions. Endogenous FilGAP colocalized with E-cadherin at adherens junctions, and depletion of FilGAP reduced the amount of E-cadherin expressed at the surface. The Rac GAP domain of FilGAP was necessary for the suppression of cell scattering induced by HGF. In agreement with this, siRNA-mediated knockdown of both Rac1 and FilGAP suppressed cell scattering induced by HGF. Forced expression of Rho kinase (ROCK, of which there are two isoforms ROCK1 and ROCK2) induced the accumulation of E-cadherin at the adherens junction, and depletion of FilGAP prevented the accumulation of E-cadherin. Moreover, wild-type FilGAP but not a non-phosphorylatable FilGAP mutant rescued the accumulation of E-cadherin at adherens junctions. These results suggest that FilGAP might regulate cell-cell adhesion through inactivation of Rac downstream of Rho-ROCK-signaling in MDCK cells.
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
DIC, 99%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Triton X-100, laboratory grade
SAFC
Sodium deoxycholate
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O