Merck
  • Ultrafast charge carrier relaxation and charge transfer processes in CdS/CdTe thin films.
Ultrafast charge carrier relaxation and charge transfer processes in CdS/CdTe thin films.
Physical chemistry chemical physics : PCCP (2015-06-03)
Bill Pandit, Ruvini Dharmadasa, I M Dharmadasa, Thad Druffel, Jinjun Liu
ABSTRACT
Ultrafast transient absorption pump-probe spectroscopy (TAPPS) has been employed to investigate charge carrier relaxation in cadmium sulfide/cadmium telluride (CdS/CdTe) nanoparticle (NP)-based thin films and electron transfer (ET) processes between CdTe and CdS. Effects of post-growth annealing treatments to ET processes have been investigated by carrying out TAPPS experiments on three CdS/CdTe samples: as deposited, heat treated, and CdCl2 treated. Clear evidence of ET process in the treated thin films has been observed by comparing transient absorption (TA) spectra of CdS/CdTe thin films to those of CdS and CdTe. Quantitative comparison between ultrafast kinetics at different probe wavelengths unravels the ET processes and enables determination of its rate constants. Implication of the photoinduced dynamics to photovoltaic devices is discussed.
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, ≥97.0%