Merck
  • Sgk1 regulates desmoglein 1 expression levels in oligodendrocytes in the mouse corpus callosum after chronic stress exposure.
Sgk1 regulates desmoglein 1 expression levels in oligodendrocytes in the mouse corpus callosum after chronic stress exposure.
Biochemical and biophysical research communications (2015-06-06)
Shingo Miyata, Keiko Yoshikawa, Manabu Taniguchi, Toshiko Ishikawa, Takashi Tanaka, Shoko Shimizu, Masaya Tohyama
ABSTRACT
Major depression, one of the most prevalent mental illnesses, is thought to be a multifactorial disease related to both genetic and environmental factors. However, the genes responsible for and the pathogenesis of major depression at the molecular level remain unclear. Recently, we reported that stressed mice with elevated plasma corticosterone levels show upregulation and activation of serum glucocorticoid-regulated kinase (Sgk1) in oligodendrocytes. Active Sgk1 causes phosphorylation of N-myc downstream-regulated gene 1 (Ndrg1), and phospho-Ndrg1 increases the expression of N-cadherin, α-catenin, and β-catenin in oligodendrocytes. This activation of the Sgk1 cascade results in morphological changes in the oligodendrocytes of nerve fiber bundles, such as those present in the corpus callosum. However, little is known about the molecular functions of the traditional and/or desmosomal cadherin superfamily in oligodendrocytes. Therefore, in this study, we aimed to elucidate the functions of the desmosomal cadherin superfamily in oligodendrocytes. Desmoglein (Dsg) 1, Dsg2, and desmocollin 1 (Dsc1) were found to be expressed in the corpus callosum of mouse brain, and the expression of a subtype of Dsg1, Dsg1c, was upregulated in oligodendrocytes after chronic stress exposure. Furthermore, Dsg1 proteins were localized around the plasma membrane regions of oligodendrocytes. A study in primary oligodendrocyte cultures also revealed that chronic upregulation of Sgk1 by dexamethasone administration is involved in upregulation of Dsg1c mRNA. These results may indicate that chronic stress induced Sgk1 activation in oligodendrocytes, which increases Dsg1 expression near the plasma membrane. Thus, Dsg1 upregulation may be implicated in the molecular mechanisms underlying the morphological changes in oligodendrocytes in response to chronic stress exposure.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SGK3, C8ORF44-SGK3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sgk2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sgk3
Sigma-Aldrich
MISSION® esiRNA, targeting human SGK2