Merck
  • Structure-based design and synthesis of phosphinate isosteres of phosphotyrosine for incorporation in Grb2-SH2 domain inhibitors. Part 1.
Structure-based design and synthesis of phosphinate isosteres of phosphotyrosine for incorporation in Grb2-SH2 domain inhibitors. Part 1.
Bioorganic & medicinal chemistry letters (2000-10-31)
P Furet, G Caravatti, A A Denholm, A Faessler, H Fretz, C García-Echeverría, B Gay, E Irving, N J Press, J Rahuel, J Schoepfer, C V Walker
ABSTRACT
Based on X-ray crystal structure information, mono charged phosphinate isosteres of phosphotyrosine have been designed and incorporated in a short inhibitory peptide sequence of the Grb2-SH2 domain. The resulting compounds, by exploiting additional interactions, inhibit binding to the Grb2-SH2 domain as potently as the corresponding doubly charged (phosphonomethyl)phenylalanine analogue.