Merck
  • Single-step, paper-based concentration and detection of a malaria biomarker.
Single-step, paper-based concentration and detection of a malaria biomarker.
Analytica chimica acta (2015-06-05)
David Y Pereira, Ricky Y T Chiu, Samantha C L Zhang, Benjamin M Wu, Daniel T Kamei
ABSTRACT
The lateral-flow immunoassay (LFA) is an inexpensive and rapid paper-based assay that can potentially detect infectious disease biomarkers in resource-poor settings. Despite its many advantages that make it suitable for point-of-care diagnosis, LFA is limited by its inferior sensitivity relative to sophisticated laboratory-based assays. Our group previously introduced the use of a micellar aqueous two-phase system (ATPS), comprised of the nonionic Triton X-114 surfactant, to concentrate biomarkers in a sample and enhance their detection with LFA. However, achieving complete phase separation and target concentration using the Triton X-114 system required many hours, and the concentrated sample needed to be manually extracted and applied to LFA. Here, we successfully integrated the concentration and detection steps into a single step that occurs entirely within a portable paper-based diagnostic strip. In a novel approach, we applied the micellar ATPS to a 3-D paper design and effectively reduced the macroscopic phase separation time from 8 h to approximately 3 min. The 3-D design was integrated with LFA to simultaneously concentrate and detect Plasmodium lactate dehydrogenase (pLDH), a malaria biomarker, in both phosphate-buffered saline and fetal bovine serum within 20 min at room temperature. Compared to a conventional LFA setup with a pLDH detection limit of 10 ng μL(-1), our single-step diagnostic successfully detected pLDH at 1.0 ng μL(-1), demonstrating a 10-fold detection limit improvement and resulting in a sensitive and user-friendly assay that can be used at the point-of-care. The integration of a micellar ATPS and LFA represents a new platform that can improve and promote the use of paper-based diagnostic assays for malaria and other diseases within resource-poor settings.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis