Merck
  • Adrenomedullin improves intestinal epithelial barrier function by downregulating myosin light chain phosphorylation in ulcerative colitis rats.
Adrenomedullin improves intestinal epithelial barrier function by downregulating myosin light chain phosphorylation in ulcerative colitis rats.
Molecular medicine reports (2015-06-06)
Zaifeng Yi, Heng Fan, Xingxing Liu, Qing Tang, Dongmei Zuo, Jia Yang
ABSTRACT
Adrenomedullin (AM) is a pivotal endogenous vasoactive peptide, which can maintain epithelial barrier function in inflammatory bowel disease. Myosin light chain kinase (MLCK)‑dependent phosphorylated myosin light chain kinase (p‑MLC) is a key regulator of intestinal barrier function. The aim of the present study was to investigate the effect and mechanism of AM on the intestinal epithelial barrier in a rat model of ulcerative colitis (UC) induced by 2,4,6‑trinitro‑benzene‑sulfonic acid (TNBS). A total of 21 male Sprague‑Dawley rats were randomly divided into the following three groups and administered different agents for 7 days: The normal group (water and saline), model group (TNBS and saline) and the AM group (TNBS and AM; 1.0 µg). The weight of rats was recorded every day. Serum tumor necrosis factor‑α (TNF‑α) and interleukin‑6 (IL‑6) levels were detected using ELISA kits. Colon tissue was collected for the assessment of histological alterations. The protein expression of MLCK, p‑MLC and zonula occludens‑1 (ZO‑1) was examined by western blot analysis. Intestinal epithelial tight junctions were examined using transmission electron microscopy. The results demonstrated that in colitis model rats, the expression of TNF‑α, IL‑6, MLCK and p‑MLC significantly increased compared with normal rats. In addition, the expression of ZO‑1 decreased (P<0.05) and intestinal epithelial cell permeability increased. Following AM administration, TNF‑α, IL‑6, MLCK and p‑MLC expression significantly decreased compared with the model rats, the expression of ZO‑1 increased (P<0.05) and intestinal epithelial cell permeability reduced. These data indicate a protective effect of AM on intestinal epithelial barrier dysfunction via suppression of inflammatory cytokines and downregulation of MLCK‑p‑MLC in TNBS‑induced UC. In conclusion, AM/MLCK‑p‑MLC may be an important signaling pathway in the occurrence and development of UC.
Sigma-Aldrich
Anti-ZO-1 Antibody, clone R40.76, clone R40.76, from rat