Merck
  • Multiphase Postmortem Computed Tomography Angiography in Pediatrics: A Case Report.
Multiphase Postmortem Computed Tomography Angiography in Pediatrics: A Case Report.
The American journal of forensic medicine and pathology (2015-08-19)
Christine Chevallier, Katarzyna Michaud, Cristian Palmiere, Leonor Alamo, Patrice Mangin, Silke Grabherr
ABSTRACT
Postmortem imaging techniques, especially postmortem computed tomography, have become integral tools in forensic investigations. Multiphase postmortem computed tomography angiography (MPMCTA) visualizes the vascular system in detail and makes it possible to evaluate the perfusion of even the smallest vessels. Although the technique has been well described for adults, no pediatric cases have been reported and no pediatric protocol has been established for this type of investigation. We present the case a 7-year-old child for which we used a previously described MPMCTA protocol and adapted values of perfusion, with the same technical equipment as for adult cases. Our main objective was to propose a perfusion protocol adapted for the investigation of infants and children. Moreover, we discuss both the difficulties that we encountered and possible ways to further improve the investigation of pediatric cases by MPMCTA.
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodine, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
SKU
Pack Size
Availability
Price
Quantity
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)