Merck
  • Vitamin A-Deficient Hosts Become Nonsymptomatic Reservoirs of Escherichia coli-Like Enteric Infections.
Vitamin A-Deficient Hosts Become Nonsymptomatic Reservoirs of Escherichia coli-Like Enteric Infections.
Infection and immunity (2015-05-13)
Kaitlin L McDaniel, Katherine H Restori, Jeffery W Dodds, Mary J Kennett, A Catharine Ross, Margherita T Cantorna
ABSTRACT
Vitamin A deficiency (A(-)) remains a public health concern in developing countries and is associated with increased susceptibility to infection. Citrobacter rodentium was used to model human Escherichia coli infections. A(-) mice developed a severe and lethal (40%) infection. Vitamin A-sufficient (A(+)) mice survived and cleared the infection by day 25. Retinoic acid treatment of A(-) mice at the peak of the infection eliminated C. rodentium within 16 days. Inflammation levels were not different between A(+) and A(-) mouse colons, although the A(-) mice were still infected at day 37. Increased mortality of A(-) mice was not due to systemic cytokine production, an inability to clear systemic C. rodentium, or increased pathogenicity. Instead, A(-) mice developed a severe gut infection with most of the A(-) mice surviving and resolving inflammation but not eliminating the infection. Improvements in vitamin A status might decrease susceptibility to enteric pathogens and prevent potential carriers from spreading infection to susceptible populations.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Nalidixic acid, ≥98%