Merck
Arginine-induced insulin secretion in endoplasmic reticulum.
Biochemical and biophysical research communications (2015-09-09)
Makoto Umeda, Masaki Hiramoto, Atsushi Watanabe, Nobuyo Tsunoda, Takeshi Imai
ABSTRACT
Arginine, a semi-essential amino acid, is known as one of the most strongest insulin secretagogues in a glucose-dependent manner, but major mechanism is unknown. Arginine induced insulin secretion in mice as well as β cell line, NIT-1, in which more than 90% of intracellular insulin is prionsulin without arginine cultivation. Arginine administration reduced prionsulin amount in 30 s, then insulin is secreted from NIT1 cells. These data indicated that the target factor(s) for arginine-induced insulin secretion located in endoplasmic reticulum (ER). We established the screening system for identifying the arginine mimetics. Brazilian propolis, not Chinese propolis, induced insulin secretion. To identify target factor(s) of arginine induced insulin secretion, our previous study was that nanobeads technology facilitated us to purify chemical-target factors. This time we chose the other way, proinsulin associating factor purification and arginine-immobilized agarose. Three proinsulin associating factors and 5 arginine interacting factors were identified. Among theses factors, Calnexin (CNX) was the only one factor, which belonged to both groups, suggesting that CNX might play a key role in arginine-induced insulin secretion in ER.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
SAFC
Iodoacetamide