Merck
Negative Skeletal Effects of Locally Produced Adiponectin.
PloS one (2015-08-01)
Marcia J Abbott, Theresa M Roth, Linh Ho, Liping Wang, Dylan O'Carroll, Robert A Nissenson
ABSTRACT
Epidemiological studies show that high circulating levels of adiponectin are associated with low bone mineral density. The effect of adiponectin on skeletal homeostasis, on osteoblasts in particular, remains controversial. We investigated this issue using mice with adipocyte-specific over-expression of adiponectin (AdTg). MicroCT and histomorphometric analysis revealed decreases (15%) in fractional bone volume in AdTg mice at the proximal tibia with no changes at the distal femur. Cortical bone thickness at mid-shafts of the tibia and at the tibiofibular junction was reduced (3-4%) in AdTg mice. Dynamic histomorphometry at the proximal tibia in AdTg mice revealed inhibition of bone formation. AdTg mice had increased numbers of adipocytes in close proximity to trabecular bone in the tibia, associated with increased adiponectin levels in tibial marrow. Treatment of BMSCs with adiponectin after initiation of osteoblastic differentiation resulted in reduced mineralized colony formation and reduced expression of mRNA of osteoblastic genes, osterix (70%), Runx2 (52%), alkaline phosphatase (72%), Col1 (74%), and osteocalcin (81%). Adiponectin treatment of differentiating osteoblasts increased expression of the osteoblast genes PPARγ (32%) and C/ebpα (55%) and increased adipocyte colony formation. These data suggest a model in which locally produced adiponectin plays a negative role in regulating skeletal homeostasis through inhibition of bone formation and by promoting an adipogenic phenotype.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Adiponectin from mouse, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)