Merck
  • The Role of Long Intergenic Noncoding RNA p21 in Vascular Endothelial Cells.
The Role of Long Intergenic Noncoding RNA p21 in Vascular Endothelial Cells.
DNA and cell biology (2015-08-15)
Chao He, Jia-Wang Ding, Song Li, Hui Wu, Yu-Rong Jiang, Wei Yang, Lin Teng, Jun Yang, Jian Yang
ABSTRACT
Large intergenic long noncoding RNA p21 (lincRNA-p21) has recently shown to play an important role in biological functions. However, the biological role of lincRNA-p21 in vascular endothelial cells remains unclear. In the present study, we investigated the role of lincRNA-p21 in vascular endothelial cells through gain- and loss-of-function studies and found that lincRNA-p21 promoted cell apoptosis and induced cell cycle progression. Furthermore, lincRNA-p21 acted as an endogenous sponge by directly binding to miR-130b and decreased miR-130b expression. In addition, miR-130b reversed the inhibitory effect of lincRNA-p21 on the growth of vascular endothelial cells. Taken together, our data highlight the pivotal role of lincRNA-p21 in the growth of vascular endothelial cells.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester, BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)