Merck
CHIP functions an E3 ubiquitin ligase of Runx1.
Biochemical and biophysical research communications (2009-06-16)
Yu Shang, Xinghui Zhao, Xialian Xu, Hong Xin, Xueni Li, Yonggong Zhai, Dacheng He, Baoqing Jia, Wei Chen, Zhijie Chang
ABSTRACT
Runx1 is a key factor in the generation and maintenance of hematopoietic stem cells. Improper expression and mutations in Runx1 are frequently implicated in human leukemia. Here, we report that CHIP, the carboxyl terminus of Hsc70-interacting protein, also named Stub1, physically interacts with Runx1 through the TPR and Charged domains in the nucleus. Over-expression of CHIP directly induced Runx1 ubiquitination and degradation through the ubiquitin-proteasome pathway. Interestingly, we found that CHIP-mediated degradation of Runx1 is independent of the molecular chaperone Hsp70/90. Taken together, we propose that CHIP serves as an E3 ubiquitin ligase that regulates Runx1 protein stability via an ubiquitination and degradation mechanism that is independent of Hsp70/90.