Merck
  • Cloning of human junctional adhesion molecule 3 (JAM3) and its identification as the JAM2 counter-receptor.
Cloning of human junctional adhesion molecule 3 (JAM3) and its identification as the JAM2 counter-receptor.
The Journal of biological chemistry (2001-10-09)
M P Arrate, J M Rodriguez, T M Tran, T A Brock, S A Cunningham
ABSTRACT
We have identified a third member of the junctional adhesion molecule (JAM) family. At the protein level JAM3 displays 36 and 32% identity to JAM2 and JAM1, respectively. The coding region is distributed over 9 exons and maps to chromosome 11q25. The gene shows widespread tissue expression with higher levels apparent in the kidney, brain, and placenta. At the cellular level we show expression of JAM3 transcript within endothelial cells. Our major finding is that JAM3 and JAM2 are binding partners. Thus, JAM3 ectodomain binds firmly to JAM2-Fc. This heterotypic interaction is maintained when JAM3-Fc is used to capture Chinese hamster ovary cells expressing full-length JAM2. In static adhesion assays we show that JAM3 is unable to bind to leukocyte cell lines. This is consistent with the lack of JAM2 expression. However, using JAM2-Fc pull-down experiments in combination with polyclonal anti-JAM3 serum, we demonstrate that JAM3 is the previously uncharacterized 43-kDa counter-receptor that mediates JAM2 adhesion to T cells. Most significantly we demonstrate up-regulation of JAM3 protein on peripheral blood lymphocytes following activation. Finally we show the utility of JAM3 ectodomain as an inhibitor of JAM2 adhesion.