Merck
  • Forced IFIT-2 expression represses LPS induced TNF-alpha expression at posttranscriptional levels.
Forced IFIT-2 expression represses LPS induced TNF-alpha expression at posttranscriptional levels.
BMC immunology (2008-12-26)
Susanne Berchtold, Birgit Manncke, Juliane Klenk, Julia Geisel, Ingo B Autenrieth, Erwin Bohn
ABSTRACT
Interferon induced tetratricopeptide repeat protein 2 (IFIT-2, P54) belongs to the type I interferon response genes and is highly induced after stimulation with LPS. The biological function of this protein is so far unclear. Previous studies indicated that IFIT-2 binds to the initiation factor subunit eIF-3c, affects translation initiation and inhibits protein synthesis. The aim of the study was to further characterize the function of IFIT-2. Stimulation of RAW264.7 macrophages with LPS or IFN-gamma leads to the expression of IFIT-2 in a type I interferon dependent manner. By using stably transfected RAW264.7 macrophages overexpressing IFIT-2 we found that IFIT-2 inhibits selectively LPS induced expression of TNF-alpha, IL-6, and MIP-2 but not of IFIT-1 or EGR-1. In IFIT-2 overexpressing cells TNF-alpha mRNA expression was lower after LPS stimulation due to reduced mRNA stability. Further experiments suggest that characteristics of the 3'UTR of transcripts discriminate whether IFIT-2 has a strong impact on protein expression or not. Our data suggest that IFIT-2 may affect selectively LPS induced protein expression probably by regulation at different posttranscriptional levels.
Sigma-Aldrich
Anti-IFIT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution