Merck
  • Comparability of higher order structure in proteins: chemometric analysis of second-derivative amide I Fourier transform infrared spectra.
Comparability of higher order structure in proteins: chemometric analysis of second-derivative amide I Fourier transform infrared spectra.
Journal of pharmaceutical sciences (2014-11-11)
Gregory Stockdale, Brian M Murphy, Jennifer D'Antonio, Mark Cornell Manning, Wasfi Al-Azzam
ABSTRACT
Comparing higher order structure (HOS) in therapeutic proteins is a significant challenge. Previously, we showed that changes in solution conditions produced detectable changes in the second-derivative amide I Fourier transform infrared (FTIR) spectra for a variety of model proteins. Those comparisons utilized vector-based approaches, such as spectral overlap and spectral correlation coefficients to quantify differences between spectra. In this study, chemometric analyses of the same data were performed, to classify samples into different groups based on the solution conditions received. The solution conditions were composed of various combinations of temperature, pH, and salt types. At first, principal component analysis (PCA) was used to visually demonstrate that FTIR spectra respond to changes in solution conditions, which, presumably indicates variations in HOS. This observed when samples from the same solution condition form clusters within a PCA score plot. The second approach, called soft independent modeling of class analogy (SIMCA), was conducted to account for the within-class experimental error for the lysozyme spectra. The DModX values, indicative of the distance of each spectra to their respective class models, was found to be a more sensitive quantitative indicator of changes in HOS, when compared with the modified area of overlap algorithm. The SIMCA approach provides a metric to determine whether new observations do, or do not belong to a particular class or group. Thus, SIMCA is the recommended approach when multiple samples from each condition are available.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SAFC
Glycine
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)