Merck
  • Monocytes regulate systemic coagulation and inflammation in abdominal sepsis.
Monocytes regulate systemic coagulation and inflammation in abdominal sepsis.
American journal of physiology. Heart and circulatory physiology (2014-12-17)
Yongzhi Wang, Oscar Ö Braun, Su Zhang, Eva Norström, Henrik Thorlacius
ABSTRACT
Abdominal sepsis is associated with significant changes in systemic inflammation and coagulation. The purpose of the present study was to examine the role of peripheral blood monocytes for systemic coagulation, including thrombin generation and consumption of coagulation factors. Abdominal sepsis was induced by cecal ligation and puncture (CLP) in C57BL/6 mice. Plasma and lung levels of IL-6 and C-X-C motif (CXC) chemokines [chemokine CXC ligand (CXCL)1, CXCL2, and CXCL5], pulmonary activity of myeloperoxidase, thrombin generation, and coagulation factors were determined 6 h after CLP induction. Administration of clodronate liposomes decreased circulating levels of monocytes by 96%. Time to peak thrombin formation was increased and peak and total thrombin generation was decreased in plasma from CLP animals. Monocyte depletion decreased time to peak formation of thrombin and increased peak and total generation of thrombin in septic animals. In addition, monocyte depletion decreased the CLP-induced increase in the levels of thrombin-antithrombin complexes in plasma. Depletion of monocytes increased plasma levels of prothrombin, factor V, factor X, and protein C in septic mice. Moreover, depletion of monocytes decreased CLP-induced levels of IL-6 and CXC chemokines in the plasma and lung by >59% and 20%, respectively. CLP-induced myeloperoxidase activity in the lung was attenuated by 44% in animals depleted of monocytes. Taken together, our findings show, for the first time, that peripheral blood monocytes regulate systemic coagulation. The results of our study improve our understanding of the pathophysiology of sepsis and encourage further attempts to target innate immune cell functions in abdominal sepsis.
Sigma-Aldrich
Xylazine, ≥99%
Ketamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Ketamine hydrochloride, solid