Merck
  • Porcine group A rotaviruses with heterogeneous VP7 and VP4 genotype combinations can be found together with enteric bacteria on Belgian swine farms.
Porcine group A rotaviruses with heterogeneous VP7 and VP4 genotype combinations can be found together with enteric bacteria on Belgian swine farms.
Veterinary microbiology (2014-05-20)
Sebastiaan Theuns, Lowiese M B Desmarets, Elisabeth Heylen, Mark Zeller, Annelike Dedeurwaerder, Inge D M Roukaerts, Marc Van Ranst, Jelle Matthijnssens, Hans J Nauwynck
ABSTRACT
Group A rotaviruses (RVA) are an important cause of diarrhea in young piglets, resulting in significant economic losses. However, the role of RVA in the etiology of piglet diarrhea on Belgian swine farms was previously unreported. In the present study, different techniques, including fast antigen detection tests, virus isolation, RT-PCR and RT-qPCR have been applied for detection of RVA in diarrheic (n=28) and asymptomatic (n=6) fecal samples collected on Belgian pig farms. RT-qPCR was shown to be most sensitive. Routine bacteriological analysis of the fecal samples showed that most diarrheic RVA positive samples were also co-infected with one or more bacterial species, such as Escherichia coli, Clostridium perfringens, Salmonella sp. and/or Brachyspira sp. Further genetic characterization of the VP7 and VP4 genes of 26 RVA strains resulted in the detection of six different G-genotypes (G2, G3, G4, G5, G9 and G11), and five different P-genotypes (P[6], P[7], P[13], P[23], P[27]), in a total of 12 different G/P combinations. A large intra-genotypic diversity was also apparent. In conclusion, results of the present study help us better understand the role of RVA in the pathogenesis of piglet diarrhea, and provide better insights into the vast genetic diversity present among circulating porcine group A rotaviruses.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Octane, reagent grade, 98%