Merck
  • Progression of muscle histopathology but not of spliceopathy in myotonic dystrophy type 2.
Progression of muscle histopathology but not of spliceopathy in myotonic dystrophy type 2.
Neuromuscular disorders : NMD (2014-08-21)
Rosanna Cardani, Marzia Giagnacovo, Giulia Rossi, Laura V Renna, Enrico Bugiardini, Chiara Pizzamiglio, Annalisa Botta, Giovanni Meola
ABSTRACT
Myotonic dystrophy type 2 (DM2) is an autosomal dominant progressive disease involving skeletal and cardiac muscle and brain. It is caused by a tetranucleotide repeat within the first intron of the CNBP gene that leads to an alteration of the alternative splicing of several genes. To understand the molecular mechanisms that play a role in DM2 progression, the evolution of skeletal muscle histopathology and biomolecular findings in successive biopsies have been studied. Biceps brachii biopsies from 5 DM2 patients who underwent two successive biopsies at different years of age have been used. Muscle histopathology has been assessed on sections immunostained with fast or slow myosin. FISH in combination with MBNL1-immunofluorescence has been performed to evaluate ribonuclear inclusion and MBNL1 foci dimensions in myonuclei. Gene and protein expression and alteration of alternative splicing of several genes have been evaluated over time. All DM2 patients examined show a worsening of muscle histopathology and an increase of foci dimensions over time. The progressive worsening of myotonia in DM2 patients may be due to the decrease of CLCN1 mRNA observed in all patients examined. However, a worsening of alternative splicing alterations has not been evidenced over time. The data obtained in this study confirm that DM2 is a slow progression disease since histological and biomolecular alterations observed in skeletal muscle are minimal even after 10-year interval. The data indicate that muscle morphological alterations evolve more rapidly over time than the molecular changes thus indicating that muscle biopsy is a more sensitive tool than biomolecular markers to assess disease progression at muscle level.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O