Merck
  • Low levels of IgG autoantibodies against the apolipoprotein B antigen p210 increases the risk of cardiovascular death after carotid endarterectomy.
Low levels of IgG autoantibodies against the apolipoprotein B antigen p210 increases the risk of cardiovascular death after carotid endarterectomy.
Atherosclerosis (2015-02-16)
Giuseppe Asciutto, Nuno V Dias, Andreas Edsfeldt, Ragnar Alm, Gunilla N Fredrikson, Isabel Gonçalves, Jan Nilsson
ABSTRACT
Autoimmune responses against oxidized-LDL (oxLDL) have been suggested to modulate inflammation in atherosclerosis. Previous studies showed an association between autoantibodies against the apolipoprotein B (apoB) p210 antigen and a lower risk of cardiovascular (CV) events. In the present study we investigated if autoantibodies against p210 at the time of carotid endarterectomy (CEA) predict risk for future CV events. Native (nat) and malondialdehyde (MDA)-modified apoB p210 autoantibodies (IgM-p210nat, IgG-p210nat, IgM-p210MDA and IgG-p210MDA) were analyzed by ELISA from plasma samples of 351 patients at the time they underwent CEA. The incidence of postoperative CV events was assessed using national registers. A total of 52 non-fatal and 15 fatal CV events were registered during the follow-up period (35.1 ± 16.7 months). Patients who suffered from a fatal CV event had significantly lower plasma levels of IgG-p210nat and IgG-p210MDA. Kaplan-Meier curves of event-free survival showed increased CV mortality in patients with levels of IgG-p210nat and IgG-p210MDA below the median (Log Rank 7.813, p .005 and 9.105, p .003 respectively). The association between low levels of p210 IgG and fatal post-operative CV events remained significant when adjusting for age, sex, total cholesterol, HDL cholesterol, smoking habits and hypertension in a Cox Proportional Hazard model (hazard ratios (HR) IgG-p210nat below median: HR 6.7 (95% C.I. 1.5-30.6, p .013) and IgG-p210MDA below median: HR 7.8 (95% C.I. 1.7-35.5, p .008). The present findings support the notion that autoantibodies against LDL antigens are involved in the atherosclerotic disease process and suggest that CEA patients with low levels of IgG-p210nat and IgG-p210MDA have an increased risk of post-operative CV death.
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents