Merck
  • Dynamic changes in sRAGE levels and relationship with cardiac function in STEMI patients.
Dynamic changes in sRAGE levels and relationship with cardiac function in STEMI patients.
Clinical biochemistry (2015-01-07)
Louise J N Jensen, Søren Lindberg, Søren Hoffmann, Allan Z Iversen, Sune H Pedersen, Rasmus Møgelvang, Søren Galatius, Allan Flyvbjerg, Jan S Jensen, Mette Bjerre
ABSTRACT
Soluble receptor of advanced glycation end-products (sRAGE) may be a predictive biomarker in coronary artery disease (CAD). Patients with acute myocardial infarction (AMI) have higher sRAGE levels compared to healthy subjects. Accordingly, the aim of this study was to investigate the dynamic changes in sRAGE levels during AMI and relationship with cardiac dysfunction. We prospectively included 80 patients with ST-elevation myocardial infarction (STEMI) treated with primary percutaneous coronary intervention (pPCI). sRAGE concentrations were measured before pPCI, immediately after pPCI and again on the first and second following days. Left ventricular ejection fraction (LVEF) was evaluated 1-3 days after the pPCI and again at a median of 7months by echocardiography, and infarct size was measured by cardiac magnetic resonance. sRAGE levels were high in the early phase of AMI; sRAGE levels significantly increased after pPCI compared with sRAGE before pPCI (median ratio: 1.25, 95% CI: 1.15-1.35, P<0.001), and the increase was observed prior to Troponin I (TnI). sRAGE levels decreased notably the first day after pPCI (median ratio: 0.34, 95% CI: 0.30-0.39, P<0.001). Peak sRAGE independently associated with long-term cardiac dysfunction estimated by LVEF (P=0.008). Furthermore, sRAGE measured after pPCI associated with infarct size (P=0.038). sRAGE levels were high in the early phase rather than in the days after AMI and pPCI. The increase in sRAGE was seen before detectable changes in TnI. In addition, sRAGE was independently associated with long-term cardiac dysfunction.
Product Description
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Europium, wire reel, 100mm, diameter 1.0mm, hard, 99.95%
Europium, wire reel, 50mm, diameter 1.0mm, hard, 99.95%