Merck
  • Modulation of STAT3 and STAT5 activity rectifies the imbalance of Th17 and Treg cells in patients with acute coronary syndrome.
Modulation of STAT3 and STAT5 activity rectifies the imbalance of Th17 and Treg cells in patients with acute coronary syndrome.
Clinical immunology (Orlando, Fla.) (2015-01-13)
Yingxia Zheng, Zhihao Wang, Lin Deng, Guanghui Zhang, Xiangliang Yuan, Liya Huang, Weiping Xu, Lisong Shen
ABSTRACT
The signal transducer and activator of transcription (STAT) activity plays an important role in the differentiation and imbalance of Th17 and Treg cells in acute coronary syndrome (ACS) patients. We determined that the basal STAT3 phosphorylation level was significantly increased and exhibited a positive relationship with Th17 cells but was negatively correlated with Treg cells in ACS patients. Opposite effects were observed for STAT5 activity. Using the pharmaceutical inhibitor TG101348 or knockdown of STAT3 reduced the number of Th17 cells while promoting the number and function of Treg cells via the Janus kinase2 (JAK2)/STAT3 pathway in ACS patients. Significantly more STAT5 bound to the Foxp3 locus when STAT3 was knocked down, and overexpression of STAT5 led to an increased number of Treg cells but a decreased number of Th17 cells in ACS patients. Our findings demonstrate that modulation of STAT3/STAT5 activity rectifies the imbalance of Th17/Treg cells in ACS patients.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O