Merck
  • A rosemary extract rich in carnosic acid selectively modulates caecum microbiota and inhibits β-glucosidase activity, altering fiber and short chain fatty acids fecal excretion in lean and obese female rats.
A rosemary extract rich in carnosic acid selectively modulates caecum microbiota and inhibits β-glucosidase activity, altering fiber and short chain fatty acids fecal excretion in lean and obese female rats.
PloS one (2014-04-16)
María Romo-Vaquero, María-Victoria Selma, Mar Larrosa, María Obiol, Rocío García-Villalba, Rocío González-Barrio, Nicolas Issaly, John Flanagan, Marc Roller, Francisco A Tomás-Barberán, María-Teresa García-Conesa
ABSTRACT
Carnosic acid (CA) and rosemary extracts (RE) show body-weight, energy metabolism and inflammation regulatory properties in animal models but the mechanisms are not yet understood. Gut microbiota plays an important role in the host metabolism and inflammatory status and is modulated by the diet. The aim of this research was to investigate whether a RE enriched in CA affected caecum microbiota composition and activity in a rat model of genetic obesity. A RE (40% CA) was administered with the diet (0.5% w/w) to lean (fa/+) and obese (fa/fa) female Zucker rats for 64 days. Changes in the microbiota composition and β-glucosidase activity in the caecum and in the levels of macronutrients and short chain fatty acids (SCFA) in feces were examined. The RE increased the Blautia coccoides and Bacteroides/Prevotella groups and reduced the Lactobacillus/Leuconostoc/Pediococccus group in both types of animals. Clostridium leptum was significantly decreased and Bifidobacterium increased only in the lean rats. β-Glucosidase activity was significantly reduced and fecal fiber excretion increased in the two genotypes. The RE also increased the main SCFA excreted in the feces of the obese rats but decreased them in the lean rats reflecting important differences in the uptake and metabolism of these molecules between the two genotypes. Our results indicate that the consumption of a RE enriched in CA modifies microbiota composition and decreases β-glucosidase activity in the caecum of female Zucker rats while it increases fiber fecal elimination. These results may contribute to explain the body weight gain reducing effects of the RE. The mutated leptin receptor of the obese animals significantly affects the microbiota composition, the SCFA fecal excretion and the host response to the RE intake.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
4-Methylpentanoic acid, ≥98%, FCC, FG
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
4-Methylvaleric acid, 99%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Carnosic acid from Rosmarinus officinalis, ≥91%, powder
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Valeric acid, analytical standard
Sigma-Aldrich
Isobutyric acid, puriss. p.a., ≥99.5%
Sigma-Aldrich
Isovaleric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isovaleric acid, natural, ≥98%, FG
Sigma-Aldrich
Isobutyric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Valeric acid, ≥99%
Sigma-Aldrich
Isovaleric acid, 99%
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)