Merck
  • Amino acid, mineral, and polyphenolic profiles of black vinegar, and its lipid lowering and antioxidant effects in vivo.
Amino acid, mineral, and polyphenolic profiles of black vinegar, and its lipid lowering and antioxidant effects in vivo.
Food chemistry (2014-08-31)
Chung-Hsi Chou, Cheng-Wei Liu, Deng-Jye Yang, Yi-Hsieng Samuel Wu, Yi-Chen Chen
ABSTRACT
Black vinegar (BV) contains abundant essential and hydrophobic amino acids, and polyphenolic contents, especially catechin and chlorogenic acid via chemical analyses. K and Mg are the major minerals in BV, and Ca, Fe, Mn, and Se are also measured. After a 9-week experiment, high-fat/cholesterol-diet (HFCD) fed hamsters had higher (p<0.05) weight gains, relative visceral-fat sizes, serum/liver lipids, and serum cardiac indices than low-fat/cholesterol diet (LFCD) fed ones, but BV supplementation decreased (p<0.05) them which may resulted from the higher (p<0.05) faecal TAG and TC contents. Serum ALT value, and hepatic thiobarbituric acid reactive substances (TBARS), and hepatic TNF-α and IL-1β contents in HFCD-fed hamsters were reduced (p<0.05) by supplementing BV due to increased (p<0.05) hepatic glutathione (GSH) and trolox equivalent antioxidant capacity (TEAC) levels, and catalase (CAT) and glutathione peroxidase (GPx) activities. Taken together, the component profiles of BV contributed the lipid lowering and antioxidant effects on HFCD fed hamsters.
Sigma-Aldrich
Hesperetin, ≥95%
Supelco
Naringin, analytical standard
Supelco
Daidzein, analytical standard
Supelco
Hesperidin, analytical standard
Supelco
Luteolin, analytical standard
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Supelco
Hesperetin, analytical standard
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Neohesperidin, ≥90% (HPLC)
Quercitrin, primary reference standard
Supelco
Myricetin, analytical standard
Luteolin, primary reference standard
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Hesperidin, European Pharmacopoeia (EP) Reference Standard
Naringin, European Pharmacopoeia (EP) Reference Standard
Hesperidin, primary reference standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Glycitein, ≥97% (HPLC)