MilliporeSigma
  • Safety and efficacy of colistin versus meropenem in the empirical treatment of ventilator-associated pneumonia as part of a macro-project funded by the Seventh Framework Program of the European Commission studying off-patent antibiotics: study protocol for a randomized controlled trial.
Safety and efficacy of colistin versus meropenem in the empirical treatment of ventilator-associated pneumonia as part of a macro-project funded by the Seventh Framework Program of the European Commission studying off-patent antibiotics: study protocol for a randomized controlled trial.
Trials (2015-04-16)
Clara Rosso-Fernández, José Garnacho-Montero, Massimo Antonelli, George Dimopoulos, José Miguel Cisneros
ABSTRACT
Ventilator-associated pneumonia (VAP) is one of the most common and severe hospital-adquired infections, and multidrugresistant gram-negative bacilli (MDR-GNB) constitute the main etiology in many countries. Inappropriate empiric antimicrobial treatment is associated with increased mortality. In this context, the empirical treatment of choice for VAP is unknown. Colistin, is now the antimicrobial with greatest in vitro activity against MDR-GNB. The MagicBullet clinical trial is an investigator-driven clinical study, funded by the Seventh Framework Program of the European Commission. This is designed as a phase IV, randomized, controlled, open label, non-inferiority and international trial to assess the safety and efficacy of colistin versus meropenem in late onset VAP. The study is conducted in a total of 32 centers in three European countries (Spain, Italy and Greece) with specific high incidences of infections caused by MDR-GNB. Patients older than 18 years who develop VAP with both clinical and radiological signs, and are on mechanical ventilation for more than 96 hours, or less than 96 hours but with previous antibiotic treatment plus one week of hospitalization, are candidates for inclusion in the study. A total sample size of 496 patients will be randomized according to a severity clinical score (at the time of VAP diagnosis in a 1:1 ratio to receive either colistin 4.5 MU as a loading dose, followed by 3 MU every eight hours (experimental arm), or meropenem 2 g every eight hours (control arm), both combined with levofloxacin. Mortality from any cause at 28 days will be considered as the main outcome. Clinical and microbiological cure will be evaluated at 72 hours, eight days, the finalization of antibiotic treatment, and 28 days of follow-up. The efficacy evaluation will be performed in every patient who receives at least one study treatment drug, and with etiologic diagnosis of VAP, intention-to-treat population and per protocol analysis will be performed. Currently, there is no study being undertaken which analyzes empiric treatment of (VAP) with a suspicion of multi-resistance. Colistin, an off-patent antibiotic commercialized for more than 60 years, could widen the antibiotic alternatives for a high-mortality illness aggravated by antibiotic resistance. This trial is registered with ClinicalTrials.gov (identifier: NCT01292031 ; registered on 29 June 2012) and EudraCT (identifier: 2010-023310-31; registered on 7 February 2011).
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Penicillamine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Meropenem, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetaminophen, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Omeprazole, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Omeprazole, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Omeprazole, solid
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard