Merck
  • Enantioselective total synthesis of (+)-gigantecin: exploiting the asymmetric glycolate aldol reaction.
Enantioselective total synthesis of (+)-gigantecin: exploiting the asymmetric glycolate aldol reaction.
Journal of the American Chemical Society (2004-10-08)
Michael T Crimmins, Jin She
ABSTRACT
The enantioselective synthesis of the potent, selective, cytotoxic, annonaceous acetogenin, (+)-gigantecin, has been completed. An asymmetric glycolate aldol serves to establish the stereocenters at C13,14 and at C21,22. A Carreira asymmetric acetylide addition is used to establish the C17 stereocenter.
Sigma-Aldrich
Sodium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 x 5 kg
Please contact Customer Service for Availability
€416.00
32 kg
Please contact Customer Service for Availability
€635.00