- Electroactive C3 symmetric discotic liquid-crystalline triindoles.
Electroactive C3 symmetric discotic liquid-crystalline triindoles.
Chemical communications (Cambridge, England) (2006-12-06)
Berta Gómez-Lor, Beatriz Alonso, Ana Omenat, José Luis Serrano
PMID17146511
ABSTRACT
Discotic liquid crystals based on triindole, a novel redox active central core, have been synthesized and their mesomorphic behaviour investigated.