- Transcription-dependent nuclear localization of DAZAP1 requires an N-terminal signal.
Transcription-dependent nuclear localization of DAZAP1 requires an N-terminal signal.
Biochemical and biophysical research communications (2012-11-01)
Yi-Tzu Lin, Wan-Ching Wen, Pauline H Yen
PMID23111326
ABSTRACT
Deleted in Azoospermia Associated Protein 1 (DAZAP1) is a ubiquitous hnRNP protein required for normal development and spermatogenesis. It resides predominantly in the nucleus and moves between the nucleus and the cytoplasm via a ZNS shuttling signal at its C-terminus. DAZAP1 accumulates in the cytoplasm when RNA polymerase II activity is inhibited by actinomycin D. Here we report the mapping of a 42-amino acid segment (N42) at the N-terminus of DAZAP1 that is both necessary and sufficient for its transcription-dependent nuclear localization. In addition, using a yeast two-hybrid system, we have identified SLIRP as a N42-binding protein which may regulate DAZAP1 subcellular localization.