Merck
  • Titrating T-cell epitopes within self-assembled vaccines optimizes CD4+ helper T cell and antibody outputs.
Titrating T-cell epitopes within self-assembled vaccines optimizes CD4+ helper T cell and antibody outputs.
Advanced healthcare materials (2014-06-14)
Rebecca R Pompano, Jianjun Chen, Emily A Verbus, Huifang Han, Arthur Fridman, Tessie McNeely, Joel H Collier, Anita S Chong
ABSTRACT
Epitope content plays a critical role in determining T-cell and antibody responses to vaccines, biomaterials, and protein therapeutics, but its effects are nonlinear and difficult to isolate. Here, molecular self-assembly is used to build a vaccine with precise control over epitope content, in order to finely tune the magnitude and phenotype of T helper and antibody responses. Self-adjuvanting peptide nanofibers are formed by co-assembling a high-affinity universal CD4+ T-cell epitope (PADRE) and a B-cell epitope from Staphylococcus aureus at specifiable concentrations. Increasing the PADRE concentration from micromolar to millimolar elicited bell-shaped dose-responses that are unique to different T-cell populations. Notably, the epitope ratios that maximize T follicular helper and antibody responses differed by an order of magnitude from those that maximized Th1 or Th2 responses. Thus, modular materials assembly provides a means of controlling epitope content and efficiently skewing the adaptive immune response in the absence of exogenous adjuvant; this approach may contribute to the development of improved vaccines and immunotherapies.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, affinity purified, lyophilized from 10 mM potassium phosphate, ≥13 U/mg protein