Merck
  • A protocol for the 0.9% saline versus Plasma-Lyte 148 for intensive care fluid therapy (SPLIT) study.
A protocol for the 0.9% saline versus Plasma-Lyte 148 for intensive care fluid therapy (SPLIT) study.
Critical care and resuscitation : journal of the Australasian Academy of Critical Care Medicine (2014-12-02)
Sumeet K Reddy, Michael J Bailey, Richard W Beasley, Rinaldo Bellomo, Seton J Henderson, Diane M Mackle, Colin J McArthur, Jan E Mehrtens, John A Myburgh, Shay P McGuinness, Alex J Psirides, Paul J Young
ABSTRACT
0.9% saline is the most commonly used intravenous (IV) fluid in the world. However, recent data raise the possibility that, compared with buffered crystalloid fluids such as Plasma-Lyte 148, the administration of 0.9% saline to intensive care unit patients might increase their risk of acute kidney injury (AKI). To describe the protocol for the 0.9% Saline v Plasma-Lyte 148 for ICU Fluid Therapy (SPLIT) study. This is a multicentre, cluster-randomised, double crossover feasibility study to be conducted in four New Zealand tertiary ICUs over a 28-week period and will enroll about 2300 participants. All ICU patients who need crystalloid IV fluid therapy (except those with established renal failure needing dialysis and those admitted to the ICU for palliative care) will be enrolled. Participating ICUs will be randomly assigned to 0.9% saline or Plasma-Lyte 148 as the routine crystalloid IV fluid, in a blinded fashion, in four alternating 7-week blocks. The primary outcome will be the proportion of patients who develop AKI in the ICU. Secondary outcomes will include the difference between the most recent serum creatinine level measured before study enrollment and the peak serum creatinine level in the ICU; use of renal replacement therapy; and ICU and in hospital mortality. All analyses will be conducted on an intention-to-treat basis. The SPLIT study started on 1 April 2014 and will provide preliminary data on the comparative effectiveness of using 0.9% saline v Plasma- Lyte 148 as the routine IV fluid therapy in ICU patients.
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)