Merck
  • Occurrence of phthalate esters in river sediments in areas with different land use patterns.
Occurrence of phthalate esters in river sediments in areas with different land use patterns.
The Science of the total environment (2014-09-14)
Jun Wang, Luji Bo, Lina Li, Dejian Wang, Gangcai Chen, Peter Christie, Ying Teng
ABSTRACT
Phthalate esters (PAEs) are widely used as plasticizers in both industrial and commercial products and have aroused considerable concern over their widespread distribution and potentially hazardous impacts on the environment. The present study investigated the distribution (concentrations at different sites) of PAEs in typical riverine sediments in southern Jiangsu Province and its relationship with the physical and chemical properties of the sediments. PAEs were detected in all 34 sediment samples analyzed, and the total concentrations of the six priority control PAEs in sediments ranged from 2.3 to 80.1 mg kg(-1) with a mean concentration of 13.2 ± 14.7 mg kg(-1). Land use significantly (p<0.05) influenced the PAE concentrations in the river sediments. The average PAE concentrations in mixed industrial and commercial districts were 27.8 ± 18.2 mg kg(-1), three and eight times higher than those in suburban areas (8.8 ± 3.7 mg kg(-1)) and an agricultural field (3.4 ± 0.9 mg kg(-1)), respectively. The concentrations of PAEs showed significant linear positive correlations with sediment organic matter and phosphorus but no significant correlation with sediment pH. Di(2-ethylhexyl) phthalate (DEHP) and di-n-butyl phthalate (DnBP) were the dominant PAE compounds present in all examined sediment samples. These PAEs have relatively high ecotoxicology, but the potential risk of their transfer to the food chain needs further study.
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%