Merck
  • The Wilms' tumor antigen is a novel target for human CD4+ regulatory T cells: implications for immunotherapy.
The Wilms' tumor antigen is a novel target for human CD4+ regulatory T cells: implications for immunotherapy.
Cancer research (2008-08-05)
Cynthia Lehe, Hazem Ghebeh, Abdullah Al-Sulaiman, Ghofran Al Qudaihi, Khaled Al-Hussein, Fahad Almohareb, Naeem Chaudhri, Fahad Alsharif, Hazza Al-Zahrani, Abdelghani Tbakhi, Mahmoud Aljurf, Said Dermime
ABSTRACT
Compelling evidences indicate a key role for regulatory T cells (T(reg)) on the host response to cancer. The Wilms' tumor antigen (WT1) is overexpressed in several human leukemias and thus considered as promising target for development of leukemia vaccine. However, recent studies indicated that the generation of effective WT1-specific cytotoxic T cells can be largely affected by the presence of T(regs). We have generated T-cell lines and clones that specifically recognized a WT1-84 (RYFKLSHLQMHSRKH) peptide in an HLA-DRB1*0402-restricted manner. Importantly, they recognized HLA-DRB1*04-matched fresh leukemic cells expressing the WT1 antigen. These clones exerted a T helper 2 cytokine profile, had a CD4(+)CD25(+)Foxp3(+)GITR(+)CD127(-) T(reg) phenotype, and significantly inhibited the proliferative activity of allogeneic T cells independently of cell contact. Priming of alloreactive T cells in the presence of T(regs) strongly inhibited the expansion of natural killer (NK), NK T, and CD8(+) T cells and had an inhibitory effect on NK/NK T cytotoxic activity but not on CD8(+) T cells. Furthermore, priming of T cells with the WT1-126 HLA-A0201-restricted peptide in the presence of T(regs) strongly inhibited the induction of anti-WT1-126 CD8(+) CTL responses as evidenced by both very low cytotoxic activity and IFN-gamma production. Moreover, these T(reg) clones specifically produced granzyme B and selectively induced apoptosis in WT1-84-pulsed autologous antigen-presenting cells but not in apoptotic-resistant DR4-matched leukemic cells. Importantly, we have also detected anti-WT1-84 interleukin-5(+)/granzyme B(+)/Foxp3(+) CD4(+) T(regs) in five of eight HLA-DR4(+) acute myeloid leukemia patients. Collectively, our in vitro and in vivo findings strongly suggest important implications for the clinical manipulation of T(regs) in cancer patients.
Sigma-Aldrich
CD4 (SP35) Rabbit Monoclonal Antibody