Merck
  • Mixed micelles of benzyldimethyltetradecylammonium chloride with tetradecyltrimethylammonium and tetradecyltriphenylphosphonium bromides: a head group contribution.
Mixed micelles of benzyldimethyltetradecylammonium chloride with tetradecyltrimethylammonium and tetradecyltriphenylphosphonium bromides: a head group contribution.
Journal of colloid and interface science (2004-02-06)
Mandeep Singh Bakshi, Ishpinder Kaur, Rohit Sood, Jasmeet Singh, Kulbir Singh, Shweta Sachar, Kanwar Jit Singh, Gurinder Kaur
ABSTRACT
Mixed micelle formation by tetradecyltrimethylammonium (TTAB) and tetradecyltriphenylphosphonium bromides (TTPB) with benzyldimethyltetradecylammonium chloride (BTDACl) was studied with the help of conductivity and Kraft point measurements. The BTDACl + TTAB mixtures showed synergistic interactions whereas those of BTDACl + TTPB indicated weak antagonistic behavior. From Kraft temperature measurements, the enthalpy of fusion (H(1)(0)) from solid hydrated BTDACl to the liquid state in the presence of TTAB or TTPB was computed. It was found that DeltaH(1)(0) was much more positive for BTDACl + TTPB than for BTDACl + TTAB mixtures.
Sigma-Aldrich
Benzyldimethyltetradecylammonium chloride dihydrate, 98%
Pricing and availability is not currently available.