Merck
  • Inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli O157:H7 by ultra high pressure and tert-butylhydroquinone combination.
Inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli O157:H7 by ultra high pressure and tert-butylhydroquinone combination.
Journal of microbiology (Seoul, Korea) (2008-07-08)
Yoon-Kyung Chung, Ahmed E Yousef
ABSTRACT
Antimicrobial efficacy of ultra-high-pressure (UHP) can be enhanced by application of additional hurdles. The objective of this study was to systematically assess the enhancement in pressure lethality by TBHQ treatment, against barotolerant strains of Escherichia coli O157:H7 and Listeria monocytogenes. Two L. monocytogenes Scott A and the barotolerant OSY-328 strain, and two E. coli O157:H7 strains, EDL-933 and its barotolerant mutant, OSY-ASM, were tested. Cell suspensions containing TBHQ (50 ppm, dissolved in dimethyl sulfoxide) were pressurized at 200 to 500 MPa (23+/-2 degrees C) for 1 min, plated on tryptose agar and enumerated the survivors. The TBHQ-UHP combination resulted in synergistic inactivation of both pathogens, with different degrees of lethality among strains. The pressure lethality threshold, for the combination treatment, was lower for E. coli O157:H7 (> or = 200 MPa) than for L. monocytogenes (> 300 MPa). E. coli O157:H7 strains were extremely sensitive to the TBHQ-UHP treatment, compared to Listeria strains. Interestingly, a control treatment involving DMSO-UHP combination consistently resulted in higher inactivation than that achieved by UHP alone, against all strains tested. However, sensitization of the pathogens to UHP by the additives (TBHQ in DMSO) was prominently greater for UHP than DMSO. Differences in sensitivities to the treatment between these two pathogens may be attributed to discrepancies in cellular structure or physiological functions.
Sigma-Aldrich
tert-Butylhydroquinone, 97%