Merck
  • Bis(isoquinoline N-oxide) pincers as a new type of metal cation dual channel fluorosensor.
Bis(isoquinoline N-oxide) pincers as a new type of metal cation dual channel fluorosensor.
Organic letters (2002-03-01)
Daniel Collado, Ezequiel Perez-Inestrosa, Rafael Suau, Jean-Pierre Desvergne, Henri Bouas-Laurent
ABSTRACT
[structure: see text] A new type of donor-spacer-acceptor podand system has been synthesized and proved as an efficient dual channel fluorosensor for Li+, Mg2+, and Ca2+. The known ability for the N-oxide function to bind Lewis acids is the key step in the appearance of a new emitting charge-transfer (CT) excited state. The occurrence of this CT state for alkaline earth (Mg2+ and Ca2+) and not for alkaline metals (Li+) provided a new type of dual channel fluorosensors.
Sigma-Aldrich
Tetraethylene glycol di(p-toluenesulfonate), 97%