Merck
  • Identification of the human YVH1 protein-tyrosine phosphatase orthologue reveals a novel zinc binding domain essential for in vivo function.
Identification of the human YVH1 protein-tyrosine phosphatase orthologue reveals a novel zinc binding domain essential for in vivo function.
The Journal of biological chemistry (1999-08-14)
M Muda, E R Manning, K Orth, J E Dixon
ABSTRACT
A human orthologue of the Saccharomyces cerevisiae YVH1 protein-tyrosine phosphatase is able to rescue the slow growth defect caused by the disruption of the S. cerevisiae YVH1 gene. The human YVH1 gene is located on chromosome 1q21-q22, which falls in a region amplified in human liposarcomas. The evolutionary conserved COOH-terminal noncatalytic domain of human YVH1 is essential for in vivo function. The cysteine-rich COOH-terminal domain is capable of coordinating 2 mol of zinc/mol of protein, defining it as a novel zinc finger domain. Human YVH1 is the first protein-tyrosine phosphatase that contains and is regulated by a zinc finger domain.